Keep my session open?
Ending In 
The session is expired
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Update to Avantor’s response to the coronavirus (COVID-19) pandemic

Morin, light brown powder C.I. 75660

Supplier: E. Becker & Co. GmbH
Warning

0215573305 0215573310
IC15573305EA 62.41 USD InStock
IC15573305 IC15573310
Morin, light brown powder C.I. 75660
Morin
Morin is a flavonoid with anti-oxidant properties.

It has been shown to protect cells against oxygen radical damage. Morin not only scavenges oxyradicals, but also moderately inhibits xanthine oxidase, a free-radical generating enzyme. At concentrations of 75-100 micromolar, morin inhibits oxidation of low density lipoprotein (LDL) by free radicals or Cu2+. Morin has been reported to inhibit rat brain phospha­tidyl­inositol­phos­phate kinase activity in vitro and in vivo.

Soluble in ethanol, methanol (0.5% w/v- clear yellow to brown solution) It is also soluble in water (0.25 mg/mL, 20 °C; 0.94 mg/mL, 100 °C), freely soluble in alcohol, slightly soluble in ether and acetic acid, and soluble in aqueous alkaline solutions.

Morin is a flavonoid with anti-oxidant properties.


Formula: C₁₅H₁₀O₇
Melting Pt: 303…304 °C
Storage Temperature: Ambient
MDL Number: MFCD00006826
CAS Number: 480-16-0
Order Now

 

ORDER

SPECIFICATIONS

RELATED INFORMATION

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your basket, enter a quantity and click Add to Basket.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty