Bovine Mucin (from Submaxillary glands), MP Biomedicals
Supplier: E. Becker & Co. GmbH
0215574280 0215574290 0215574291
IC15574280EA 164.94 USD InStock
IC15574280 IC15574290 IC15574291
Bovine Mucin (from Submaxillary glands), MP Biomedicals
Proteins and Peptides
Typically prepared as a suspension in water adjusted to pH 5.0 with acetic acid. A 1% w/v solution will appear turbid.
Mucin is a high molecular weight glycoprotein that is the major constituent of saliva, gastrointestinal juices, mucos and other secretions. Mucin exists as an aggregated complex in excess of 106 molecular weight. It is a rich source of O-linked oligosaccharide chains.
Albuminoid substance in mucus which gives it its ropy consistency.
Order Now
ORDER
SPECIFICATIONS
RELATED INFORMATION
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your basket, enter a quantity and click Add to Basket.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty