Keep my session open?
Ending In 
The session is expired
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Update to Avantor’s response to the coronavirus (COVID-19) pandemic

Phe-Arg β-naphthylamide dihydrochloride, powder

Supplier: E. Becker & Co. GmbH

0215612810 0215612825 0215612850
IC15612810EA 95.93 USD InStock
IC15612810 IC15612825 IC15612850
Phe-Arg β-naphthylamide dihydrochloride, powder
Phe-Arg beta-naphthylamide dihydrochloride
Soluble in methanol (50 mg/mL - clear, colorless solution). Soluble in water (50 mg/mL).


Formula: C₂₅H₃₂Cl₂N₆O₂
MDL Number: MFCD00058046
CAS Number: 100929-99-5
https://qa.digitalassets.avantorsciences.com/adaptivemedia/rendition?id=240fe04c141545c21b0edca56959e7bb30006de5&vid=240fe04c141545c21b0edca56959e7bb30006de5&prid=web&clid=SAPDAM
Order Now

 

ORDER

SPECIFICATIONS

RELATED INFORMATION

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your basket, enter a quantity and click Add to Basket.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty