Keep my session open?
Ending In 
The session is expired
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Update to Avantor’s response to the coronavirus (COVID-19) pandemic

Diatomaceous earth, white powder, non-washed

Supplier: E. Becker & Co. GmbH
Warning

0215760691 0215760694
IC15760691EA 142.78 USD InStock
IC15760691 IC15760694
Diatomaceous earth, white powder, non-washed
Diatomaceous earth
Diatomaceous earth, non-washed is used as a filtration aid. It is suitable for most routine filtrations. Highly Insoluble.

Diatomaceous earth, non-washed is used as a filtration aid. It is suitable for most routine filtrations.


Formula: SiO₂
Storage Temperature: Ambient
MDL Number: MFCD00132803
CAS Number: 61790-53-2
https://qa.digitalassets.avantorsciences.com/adaptivemedia/rendition?id=782afd01e40fa7254a5017cfad511fbd830d219a&vid=782afd01e40fa7254a5017cfad511fbd830d219a&prid=web&clid=SAPDAM
Order Now

 

ORDER

SPECIFICATIONS

RELATED INFORMATION

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your basket, enter a quantity and click Add to Basket.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty