Keep my session open?
Ending In 
The session is expired
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Update to Avantor’s response to the coronavirus (COVID-19) pandemic

Conduritol B epoxide ≥97%, white powder

Supplier: E. Becker & Co. GmbH

0215889305 0215889325
IC15889305EA 209.16 USD InStock
IC15889305 IC15889325
Conduritol B epoxide ≥97%, white powder
Conduritol B epoxide
Conduritol B Epoxide (CBE) is a potent, irreversible inhibitor of various plant β-glucosidases and mammalian glucocerebrosidase, specific covalent inhibitor of β-glucosidase which results in the accumulation of glucocerebroside.

  • β-Glucosidase inhibitor

In vitro exposure of cells to CBE has been reported to lead to up-regulation of CD1d (cluster of differentiation 1d) expression by THP-1 cells, which is related to an increase in mRNA expression. Has been employed in in vitro and animal models for Gaucher's disease. Treatment of murine peritoneal macrophages with this inhibitor does not seem to affect lysozyme enzyme release to the medium, cell viability, or levels of intracellular lysosomal enzymes, other than the beta-glucosidase activity.

Soluble in water (25 mg/mL) and DMSO; slightly soluble in methanol. Store solutions aliquoted at -20 °C for up to 3 months.

In vitro exposure of cells to CBE has been reported to lead to up-regulation of CD1d (cluster of differentiation 1d) expression by THP-1 cells, which is related to an increase in mRNA expression. Has been employed in in vitro and animal models for Gaucher's disease. Treatment of murine peritoneal macrophages with this inhibitor does not seem to affect lysozyme enzyme release to the medium, cell viability, or levels of intracellular lysosomal enzymes, other than & beta-glucosidase activity.


Formula: C₆H₁₀O₅
Melting Pt: 159…161 °C (lit.)
Storage Temperature: Freezer
MDL Number: MFCD00077326
CAS Number: 6090-95-5
https://qa.digitalassets.avantorsciences.com/adaptivemedia/rendition?id=e937675210d07b676bfbf065ba5a6efb70864e74&vid=e937675210d07b676bfbf065ba5a6efb70864e74&prid=web&clid=SAPDAM
Order Now
Specification Test Results
Identity test Passes
Purity ≥97%

 

ORDER

SPECIFICATIONS

RELATED INFORMATION

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your basket, enter a quantity and click Add to Basket.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty