Keep my session open?
Ending In 
The session is expired
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Update to Avantor’s response to the coronavirus (COVID-19) pandemic

Tannic acid, brown powder

Supplier: E. Becker & Co. GmbH

0219027583 0219027590 0219027591
IC19027583EA 107.83 USD InStock
IC19027583 IC19027590 IC19027591
Tannic acid, brown powder
Tannic acid
Tannic Acid is used for enzyme immobilization and protein adsorption

Soluble in water (2 g/10 ml - clear solution), glycerol, alcohol, acetone. Practically insoluble in benzene, chloroform, ether, petroleum ether, carbondisulphide, carbon tetrachloride. Also soluble in ethanol (100 mg/ml - yellow to brown solution).

Tannic Acid is used for enzyme immobilization and protein adsorption.

Precaución: Research Use Only (RUO).


Formula: C₇₆H₅₂O₄₆
MW: 1701.2 g/mol
Flash Pt: 199
Storage Temperature: Ambient
MDL Number: MFCD00066397
CAS Number: 1401-55-4
Merck Index: 12,09221
Order Now
Specification Test Results
Identity Test Passes
Moisture ≤12%
FTIR Conforms to Standard

 

ORDER

SPECIFICATIONS

RELATED INFORMATION

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your basket, enter a quantity and click Add to Basket.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty