Keep my session open?
Ending In 
The session is expired
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Update to Avantor’s response to the coronavirus (COVID-19) pandemic

Estradiol benzoate, powder

Supplier: E. Becker & Co. GmbH
Danger

0219041101 0219041105
IC19041101EA 107.6 USD InStock
IC19041101 IC19041105
Estradiol benzoate, powder
Estradiol benzoate
Used in a study of general steroid metabolism and clearing, as groundwork for anabolic steroid screening in athletes.

Soluble in alcohol, acetone, dioxane, chloroform(50 mg/ml); slightly soluble in ether, vegetable oils.

Used in a study of general steroid metabolism and clearing, as groundwork for anabolic steroid screening in athletes.


Formula: C₂₅H₂₈O₃
MW: 376.496 g/mol
Storage Temperature: Ambient
MDL Number: MFCD00003692
CAS Number: 50-50-0
Order Now
Specification Test Results
Identity Test Passes
FTIR Conforms to Standard
Melting Point 190 - 200°C
Optical Rotation (c=1, CHCl3) +60 ± 3°

 

ORDER

SPECIFICATIONS

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your basket, enter a quantity and click Add to Basket.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty