Keep my session open?
Ending In 
The session is expired
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Update to Avantor’s response to the coronavirus (COVID-19) pandemic

Erythrosin B disodium salt, red powder

Supplier: E. Becker & Co. GmbH
Warning

0219044910 0219044925
IC19044910EA 57.64 USD InStock
IC19044910 IC19044925
Erythrosin B disodium salt, red powder
Erythrosin B disodium salt
A red, anionic xanthene dye, has been used as a contrast or counter stain to hematoxylin, crystal violet and other nuclear stains, in both plant and animal tissue.
Erythrosin B Sodium Salt is used as a counter stain to hematoxylin, crystal violet and other nuclear stains, in both plant and animal tissue. Erythosin B dye solution in 0.9% saline is made for intravenous injection to induce occlusion.

  • Concentration: 0.91
  • Soluble in water (20 mg/mL) or ethanol (20 mg/mL)
  • Room Temperature, desiccate

A red, anionic xanthene dye, has been used as a contrast or counter stain to hematoxylin, crystal violet and other nuclear stains, in both plant and animal tissue.


Formula: C₂₀H₆I₄Na₂O₅
MW: 879.86 g/mol
Storage Temperature: Ambient
MDL Number: MFCD00144257
CAS Number: 16423-68-0
https://qa.digitalassets.avantorsciences.com/adaptivemedia/rendition?id=83796b79e6f5e51ca5ad8a27a1134204f508be73&vid=83796b79e6f5e51ca5ad8a27a1134204f508be73&prid=web&clid=SAPDAM
Order Now
Specification Test Results
Identity Test Passes
UV/VIS: l max (water) 524-528nm

 

ORDER

SPECIFICATIONS

RELATED INFORMATION

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your basket, enter a quantity and click Add to Basket.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty