Keep my session open?
Ending In 
The session is expired
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Update to Avantor’s response to the coronavirus (COVID-19) pandemic

Vincristine sulfate, white powder

Supplier: E. Becker & Co. GmbH
Danger

0219068701 0219068725
IC19068701EA 128.16 USD InStock
IC19068701 IC19068725
Vincristine sulfate, white powder
Vincristine sulphate
Vincristine Sulfate, a dimeric Vinca alkaloid. Experiments have reported Vincristine Sulfate to demonstrate a capacity to suppress growth of proliferating cells through marked apoptosis. The cell death caused by this agent seems to result in a sustained accumulation of endogenous ceramide levels. Ceramide, has been proposed as a lipid second messenger with specific antiproliferative mediating responses.

Vincristine Sulfate is used to treat acute leukaemia, malignant lymphoma, Hodgkin′s disease, acute erythraemia, acute panmyelosis, breast cancer, Kaposi’s sarcoma and testicular cancer. It is widely used in cancer research. It is used to study multi-drug resistance. It is used to inhibit cell cycle progression at M-phase and to inhibit monoamine oxidase (MAO).

Vincristine is a plant alkaloid that inhibits microtubule assembly by binding tubulin and inducing coiled spiral aggregate formation. It arrests cell cycle in G2/M-phase by blocking mitotic spindle formation. Vincristine triggers Raf-1 activation, phosphorylation of bcl-2-family proteins, induction of p53 expression, and apoptosis in several tumor cell lines. It inhibits VEGF production in leukemia cell lines. It is a substrate for Pgp and CYP3A4. MRP1 transports vincristine in an ATP- and GSH-dependent manner. Vincristine has some immunosuppressant effect. Vincristine may also interfere with amino acid, cyclic AMP, and glutathione metabolism, calmodulin-dependent Ca2+-transport, cellular respiration, and nucleic acid and lipid biosynthesis.

Soluble in water (50% w/v); soluble in methanol; moderately soluble in chloroform; very slightly soluble in ethanol; insoluble in diethyl ether.

Vincristine Sulfate is used to treat acute leukaemia, malignant lymphoma, Hodgkin′s disease, acute erythraemia, acute panmyelosis, breast cancer, Kaposi′s sarcoma and testicular cancer. It is widely used in cancer research. It is used to study multi-drug resistance. It is used to inhibit cell cycle progression at M-phase and to inhibit monoamine oxidase (MAO).


Formula: C₄₆H₅₈N₄O₁₄S
Melting Pt: 273…281 °C (lit.)
MDL Number: MFCD00084729
CAS Number: 2068-78-2
UN: 2811
ADR: 6.1,II
Order Now

 

ORDER

SPECIFICATIONS

RELATED INFORMATION

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your basket, enter a quantity and click Add to Basket.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty