Human Albumin, MP Biomedicals
Supplier: E. Becker & Co. GmbH
02191349.1 02191349.2 0219134901 0219134905
IC191349.1EA 99.52 USD InStock
IC191349.1 IC191349.2 IC19134901 IC19134905
Human Albumin, MP Biomedicals
Proteins and Peptides
Soluble in water.
Each donor unit used in the manufacture of this product was tested by an FDA approved method for the presence of antibody to HIV 1/2, antibody to HCV, HIV-Ag, as well as for the Hepatitis B Surface Antigen (HBsAg), and found to be negative (not repeatably reactive).
The main biological function of albumin is to regulate the colloidal osmotic pressure of blood. Human and bovine albumins contain 16% nitrogen and are often used as standards in protein calibration studies. Due to their free hydrophobic region fatty acid free albumins are used to solubilize lipids in tissue culture, and are also used as blocking agents in Western blots or ELISA applications. Globulin free albumins are suitable for use in applications where no other proteins should be present (e.g., electrophoresis).
Serum albumin functions as a carrier protein for steroids, fatty acids, and thyroid hormones, and is vital in regulating the colloidal osmotic pressures of blood. Albumin is also seen to bind to exogenous substances, particularly drugs (e.g., ibuprofen, warfarin), and strongly influence their pharmacokinetics. Oxidative stress leading to changes in the redox state of albumin has widely varied effects on its physiological function.
Order Now
ORDER
SPECIFICATIONS
RELATED INFORMATION
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your basket, enter a quantity and click Add to Basket.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty