Keep my session open?
Ending In 
The session is expired
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Update to Avantor’s response to the coronavirus (COVID-19) pandemic

Staurosporine ≥98%

Supplier: E. Becker & Co. GmbH

0219140080 0219140090 0219140091
IC19140080EA 277.57 USD InStock
IC19140080 IC19140090 IC19140091
Staurosporine ≥98%
Staurosporine
Staurosporine is provided in a lyophilised form. It may be reconstituted by dissolving the lyophilized reagent in DMSO (5 mg/mL-clear, colorless solution), DMF, ethyl acetate (5 mg/mL-clear colorless solution) or methanol. Stock solutions should be further diluted with aqueous buffers just prior to use. Stability depends upon the quality of solvent used. The compound should be stored at -20 °C in the dark.

Potent inhibitor of phospholipid/calcium-dependent protein kinase. Inhibits the upregulation of VEGF expression in tumor cells.


Formula: C₂₈H₂₆N₄O₃
MDL Number: MFCD18252446
CAS Number: 62996-74-1
Order Now

 

ORDER

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your basket, enter a quantity and click Add to Basket.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty