Keep my session open?
Ending In 
The session is expired
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Update to Avantor’s response to the coronavirus (COVID-19) pandemic

Ammonium sulfate ≥99%, powder ACS

Supplier: E. Becker & Co. GmbH

02191409.5 0219140901 0219140905
IC191409.5EA 81.17 USD InStock
IC191409.5 IC19140901 IC19140905
Ammonium sulfate ≥99%, powder ACS
Ammonium sulphate
Ammonium Sulfate may be used for the precipitation or fractionation of proteins or for purification of antibodies. Useful for crystallographic analysis of nucleic acids and proteins. The use of ammonium sulfate in long PCR buffer, in PCR lysis solution, and in second strand cDNA synthesis has been reported.

Soluble freely in water; insoluble in alcohol, acetone.

Precaución: Research Use Only (RUO).


Formula: (NH₄)₂SO₄
Storage Temperature: Ambient
MDL Number: MFCD00003391
CAS Number: 7783-20-2
Order Now
Specification Test Results
Assay ≥99%
pH (5% aq soln) 5 - 6
Insoluble Matter ≤0.005%
Residue on Ignition ≤0,005%
Chloride (Cl) ≤5 ppm
Nitrate (NO₃) ≤0,001%
Phosphate (PO4) ≤5 ppm
Heavy Metals (as Pb) ≤5 ppm
Iron (Fe) ≤5 ppm

 

ORDER

SPECIFICATIONS

RELATED INFORMATION

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your basket, enter a quantity and click Add to Basket.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty