Keep my session open?
Ending In 
The session is expired
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Update to Avantor’s response to the coronavirus (COVID-19) pandemic

DNA sodium salt (Deoxyribonucleic acid sodium salt) (from calf thymus)

Supplier: E. Becker & Co. GmbH

0219512990 0219512991
IC19512990EA 311.99 USD InStock
IC19512990 IC19512991
DNA sodium salt (Deoxyribonucleic acid sodium salt) (from calf thymus)
DNA sodium salt (Deoxyribonucleic acid sodium salt)
Polynucleotide, carrier of genetic material.

  • White Fibrous Solid
  • Soluble in water or aqueous buffers

The addition of SDS or similar detergent may help solubilize DNA; however, detergents tend to denature the DNA into shorter strands.

Calf thymus DNA (CT-DNA) is a natural DNA widely used in studies of DNA binding anticancer agents and DNA binding agents that modulate DNA structure and function. Calf thymus DNA is also used in physicochemical studies of DNA behavior in solution.

Synonyms: DNA sodium salt from calf thymus; Thymonucleic acid sodium salt; ctDNA


Storage Temperature: Refrigerator
MDL Number: MFCD00130921
CAS Number: 9007-49-2
Order Now
Specification Test Results
Idendity Passes Test
Source Calf Thymus

 

ORDER

SPECIFICATIONS

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your basket, enter a quantity and click Add to Basket.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty