Keep my session open?
Ending In 
The session is expired
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Update to Avantor’s response to the coronavirus (COVID-19) pandemic

Primuline sodium salt, powder C.I. 49000

Supplier: E. Becker & Co. GmbH

0219545405 0219545425 0219545480
IC19545405EA 215.29 USD InStock
IC19545405 IC19545425 IC19545480
Primuline sodium salt, powder C.I. 49000
Primuline sodium salt
Primuline is a dye containing the benzothiazole ring system. The primulines are considered derivatives of dehydrothiotoluidine (aminobenzenyltoluylmercaptan). It is pale yellow in solution under visible light and is blue fluorescent under ultraviolet.

Primulin is used in retrograde double labeling procedure for studying axonal branching.

Moderately soluble in water. Soluble in methanol(1 mg/mL): yellow to yellow-green solution.

Primuline is a dye containing the benzothiazole ring system. The primulines are considered derivatives of dehydrothiotoluidine (aminobenzenyltoluylmercaptan). It is pale yellow in solution under visible light and is blue fluorescent under ultraviolet.


Formula: C₂₁H₁₄N₃O₃S₃Na
MW: 475.54 g/mol
MDL Number: MFCD00005798
CAS Number: 8064-60-6
https://qa.digitalassets.avantorsciences.com/adaptivemedia/rendition?id=36f9534329b18223ec6dd34c9614eb1294f6deba&vid=36f9534329b18223ec6dd34c9614eb1294f6deba&prid=web&clid=SAPDAM
Order Now

 

ORDER

SPECIFICATIONS

RELATED INFORMATION

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your basket, enter a quantity and click Add to Basket.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty