Keep my session open?
Ending In 
The session is expired
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Update to Avantor’s response to the coronavirus (COVID-19) pandemic

Ristocetin A ≥90%, powder

Supplier: E. Becker & Co. GmbH

0219548905 0219548925
IC19548905EA 94.44 USD InStock
IC19548905 IC19548925
Ristocetin A ≥90%, powder
Ristocetin A
Ristocetin A is an antibiotic that belongs to the class III glycopeptides. The compound has activity against gram-positive and gram-negative bacteria, and can be used in chromatography. Ristocetin A is structurally related to the glycopeptide antibiotics Teicoplanin and vancomycin.

Ristocetin A has platelet aggregating activity and has been used in platelet aggregation studies. Studies have demonstrated that Ristocentin A dimerizes and creates two binding sites for cell wall peptides.

Ristocetin activates platelets. Ristocetin-induced platelet agglutination (RIPA) is decreased in patients undergoing chronic hemodialysis. Together with increased plasma glycocalicin levels (a product of enzymatic cleavage of GPIb), RIPA may contribute to diminished platelet adhesion to vascular subendothelium and increased bleeding associated with uremia. Addition of ristocetin and vWf caused specific agglutination of rGPIbα-liposomes suporting the potential use of rGPIbα-liposomes to enhance platelet function in vivo and support hemostasis in thrombocytopenic individuals.

Soluble in 1 N HCl (50 mg/mL - clear and colorless to pale yellow); soluble in water (clear 5 mg+ 0.25 mL, amber solution).

Ristocetin A is an antibiotic that belongs to the class III glycopeptides. The compound has activity against gram-positive and gram-negative bacteria, and can be used in chromatography. Ristocetin A is structurally related to the glycopeptide antibiotics Teicoplanin and vancomycin.


Formula: C₉₅H₁₁₀N₈O₄₄
CAS Number: 1404-55-3
https://qa.digitalassets.avantorsciences.com/adaptivemedia/rendition?id=8958067391cf874a8b0c33b569659a8f53734467&vid=8958067391cf874a8b0c33b569659a8f53734467&prid=web&clid=SAPDAM
Order Now
Specification Test Results
Identity Test Passes
Potency (µg/mg) Report Result
Purity ≥90% Ristocetin A
Ristocetin B Sulfate Report Result

 

ORDER

SPECIFICATIONS

RELATED INFORMATION

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your basket, enter a quantity and click Add to Basket.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty