Keep my session open?
Ending In 
The session is expired
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Update to Avantor’s response to the coronavirus (COVID-19) pandemic

GHS Labels "Explosive Picto", Brady

Supplier: Brady Worldwide
121188 121201 121202
89234-534EA 43.47 USD InStock
89234-534 89234-560 89234-562
GHS Labels "Explosive Picto", Brady
Labels
Pre-printed GHS pictogram die cut labels on a card.

  • Color: Black/red on white

Order Now
 

ORDER

SPECIFICATIONS

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your basket, enter a quantity and click Add to Basket.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty