Glass Microfiber Filter Paper, Grade 121, Ahlstrom
Supplier: Ahlstrom
1210-0240 1210-0470 1210-0550 1210-0425 1210-0700 1210-0900 1210-0210 1210-0250 1210-0370 1210-1100 1210-1250 1210-1500
38010-418BX 78.57 USD InStock
38010-418 38010-429 38010-436 47745-790 47745-792 47745-794 47749-532 47749-534 47749-536 47749-538 47749-540 47749-542
Glass Microfiber Filter Paper, Grade 121, Ahlstrom
Filters Glass Fiber Filters
Grade 121 is a very thick binder-free microfiber glass filter paper that offers finer particle retention than Ahlstrom 111 with higher wet strength and loading capacity.
Order Now
ORDER
SPECIFICATIONS
RELATED INFORMATION
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your basket, enter a quantity and click Add to Basket.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty