Keep my session open?
Ending In 
The session is expired
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Update to Avantor’s response to the coronavirus (COVID-19) pandemic

Benzyl acetate for synthesis, Sigma-Aldrich®

Supplier: EMD MILLIPORE - AA – CORP

M-Clarity™ quality level = MQ200

8.03181.0100 8.03181.1000
EM8.03181.0100CS 687 USD InStock
EM8.03181.0100 EM8.03181.1000
Benzyl acetate for synthesis, Sigma-Aldrich®
Benzyl acetate


Formula: C₉H₁₀O₂
MW: 150.18 g/mol
Boiling Pt: 206
Melting Pt: -51
Flash Pt: 95
MDL Number: MFCD00008712
CAS Number: 140-11-4
Merck Index: 13,01125
Order Now
Specification Test Results
Solubility (20 °C) slightly soluble
Melting Point -51 °C
Molar Mass 150.17 g/mol
Boiling Point 205 - 207 °C
Vapor Pressure 1 hPa (45 °C)
Flash Point 95 °C
Refractive Index 1.5006 (20 °C, 589 nm)
Explosion Limit 0.9 - 8.4 %(V)
Density 1.06 g/cm3 (20 °C)
Ignition Point 460 °C
Synonyms Acetic acid benzyl ester
Hill Formula C9H10O2
Chemical Formula CH3COOCH2C6H5

 

ORDER

SPECIFICATIONS

RELATED INFORMATION

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your basket, enter a quantity and click Add to Basket.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty