Keep my session open?
Ending In 
The session is expired
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Update to Avantor’s response to the coronavirus (COVID-19) pandemic

2,5-Xylenol for synthesis, Sigma-Aldrich®

Supplier: EMD MILLIPORE - AA – CORP
Danger

M-Clarity™ quality level = MQ200

8.03771.0005 8.03771.0250
EM8.03771.0005CS 38533 USD InStock
EM8.03771.0005 EM8.03771.0250
2,5-Xylenol for synthesis, Sigma-Aldrich®
2,5-Xylenol


Formula: C₈H₁₀O
MDL Number: MFCD00002237
CAS Number: 95-87-4
UN: 2261
ADR: 6.1,II
Order Now
Specification Test Results
Solubility (20 °C) slightly soluble
Melting Point 75 °C
Molar Mass 122.16 g/mol
Boiling Point 211 °C
Vapor Pressure 0.208 hPa (25 °C)
Flash Point 80 - 95 °C
Explosion Limit 1.4 %(V)
Density 0.9 g/cm3 (75 °C)
Synonyms 2,5-DMP, p-Xylenol
Hill Formula C8H10O
Chemical Formula 2,5-(CH3)2C6H3OH

 

ORDER

SPECIFICATIONS

RELATED INFORMATION

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your basket, enter a quantity and click Add to Basket.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty