Keep my session open?
Ending In 
The session is expired
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Update to Avantor’s response to the coronavirus (COVID-19) pandemic

Methylammonium chloride for synthesis, Sigma-Aldrich®

Supplier: EMD MILLIPORE - AA – CORP

M-Clarity™ quality level = MQ200

8.06020.0005 8.06020.0250
EM8.06020.0005CS 29835 USD InStock
EM8.06020.0005 EM8.06020.0250
Methylammonium chloride for synthesis, Sigma-Aldrich®
Methylammonium chloride


Formula: CH₆ClN
MDL Number: MFCD00012849
CAS Number: 593-51-1
Order Now
Specification Test Results
Solubility (20 °C) soluble
Melting Point 228 - 233 °C
Molar Mass 67.52 g/mol
Bulk Density 1100 kg/m3
Boiling Point 225 - 230 °C (20 hPa)
pH 5 - 7 (10 g/l, H2O, 20 °C)
Hill Formula CH6ClN
Chemical Formula CH3NH3Cl

 

ORDER

SPECIFICATIONS

RELATED INFORMATION

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your basket, enter a quantity and click Add to Basket.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty