Keep my session open?
Ending In 
The session is expired
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Update to Avantor’s response to the coronavirus (COVID-19) pandemic

Polyphosphoric acid for synthesis, Sigma-Aldrich®

Supplier: EMD MILLIPORE - AA – CORP
Danger

M-Clarity™ quality level = MQ200

8.07471.0100 8.07471.0500 8.07471.2500
EM8.07471.0100CS 9731 USD InStock
EM8.07471.0100 EM8.07471.0500 EM8.07471.2500
Polyphosphoric acid for synthesis, Sigma-Aldrich®
Polyphosphoric acid


Formula: H(PO₃H)nOH
MDL Number: MFCD00084480
CAS Number: 8017-16-1
UN: 3264
ADR: 8 II
Order Now
Specification Test Results
Solubility (20 °C) soluble,Hydrolysis
Melting Point -20 °C
Boiling Point 530 °C (1013 hPa)
Vapor Pressure 2 hPa (20 °C)
Density 2.06 g/cm3 (20 °C)
pH (H2O, 20 °C) acidic,Hydrolysis
Synonyms PPA
Hill Formula HO[P(OH)(O)O](n)H

 

ORDER

SPECIFICATIONS

RELATED INFORMATION

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your basket, enter a quantity and click Add to Basket.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty