Keep my session open?
Ending In 
The session is expired
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Update to Avantor’s response to the coronavirus (COVID-19) pandemic

Methyl toluene-4-sulphonate for synthesis, Sigma-Aldrich®

Supplier: EMD MILLIPORE - AA – CORP
Danger

M-Clarity™ quality level = MQ200

8.08333.0500 8.08333.2500
EM8.08333.0500CS 860 USD InStock
EM8.08333.0500 EM8.08333.2500
Methyl toluene-4-sulphonate for synthesis, Sigma-Aldrich®
Methyl toluene-4-sulphonate


Formula: C₈H₁₀O₃S
MW: 186.23 g/mol
Boiling Pt: 292
Flash Pt: 130
MDL Number: MFCD00008417
CAS Number: 80-48-8
UN: 3261
ADR: 8 II
Order Now
Specification Test Results
Solubility (20 °C) insoluble
Melting Point 24 - 27 °C
Molar Mass 186.23 g/mol
Boiling Point 292 °C (1013 hPa)
Flash Point 130 °C
Density 1.23 g/cm3 (20 °C)
Synonyms p-Toluenesulfonic acid methyl ester
Hill Formula C8H10O3S
Chemical Formula 4-(CH3)C6H4SO3CH3

 

ORDER

SPECIFICATIONS

RELATED INFORMATION

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your basket, enter a quantity and click Add to Basket.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty