Keep my session open?
Ending In 
The session is expired
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Update to Avantor’s response to the coronavirus (COVID-19) pandemic

o-Cresol for synthesis, Sigma-Aldrich®

Supplier: EMD MILLIPORE - AA – CORP
Danger

M-Clarity™ quality level = MQ200

8.09692.0100 8.09692.0500 8.09692.1000 8.09692.2500
EM8.09692.0100CS 697 USD InStock
EM8.09692.0100 EM8.09692.0500 EM8.09692.1000 EM8.09692.2500
o-Cresol for synthesis, Sigma-Aldrich®
o-Cresol


Formula: C₇H₈O
MDL Number: MFCD00002226
CAS Number: 95-48-7
UN: 3455
ADR: 6.1 (8),II
Order Now
Specification Test Results
Solubility 20 g/l (20 °C)
Melting Point 31 °C
Molar Mass 108.14 g/mol
Boiling Point 191 - 192 °C (1013 hPa)
Vapor Pressure 0.24 hPa (20 °C)
Flash Point 81 °C
Refractive Index 1.553 (20 °C, 589 nm)
Explosion Limit 1.3 %(V)
Density 1.046 g/cm3 (20 °C)
pH 4.8 (20 g/l, H2O, 20 °C)
Ignition Point 555 °C
Synonyms 2-Hydroxytoluene, 2-Methylphenol
Hill Formula C7H8O

 

ORDER

SPECIFICATIONS

RELATED INFORMATION

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your basket, enter a quantity and click Add to Basket.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty