Keep my session open?
Ending In 
The session is expired
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Update to Avantor’s response to the coronavirus (COVID-19) pandemic

(1S)-(-)-3-Oxocamphorsulfonylimine for synthesis, Sigma-Aldrich®

Supplier: EMD MILLIPORE - AA – CORP

M-Clarity™ quality level = MQ200

8.14388.0001 8.14388.0005
EM8.14388.0001CS 4360 USD InStock
EM8.14388.0001 EM8.14388.0005
(1S)-(-)-3-Oxocamphorsulfonylimine for synthesis, Sigma-Aldrich®
(1S)-(-)-3-Oxocamphorsulfonylimine


Formula: C₁₀H₁₃NO₃S
MDL Number: MFCD00066268
CAS Number: 119106-38-6
Order Now
Specification Test Results
Solubility (20 °C) insoluble
Melting Point 195 - 198 °C
Molar Mass 227.28 g/mol
Hill Formula C10H13NO3S

 

ORDER

SPECIFICATIONS

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your basket, enter a quantity and click Add to Basket.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty