Keep my session open?
Ending In 
The session is expired
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Update to Avantor’s response to the coronavirus (COVID-19) pandemic

Allyl cyanoacetate for synthesis, Sigma-Aldrich®

Supplier: EMD MILLIPORE - AA – CORP
Danger

M-Clarity™ quality level = MQ200

8.18559.0050 8.18559.1000
EM8.18559.0050CS 18859 USD InStock
EM8.18559.0050 EM8.18559.1000
Allyl cyanoacetate for synthesis, Sigma-Aldrich®
Allyl cyanoacetate


Formula: C₆H₇NO₂
MDL Number: MFCD00013817
CAS Number: 13361-32-5
UN: 3276
ADR: 6.1,III
Order Now
Specification Test Results
Solubility (20 °C) insoluble
Melting Point -40.0 °C
Molar Mass 125.13 g/mol
Boiling Point 89 - 90 °C (5 hPa)
Vapor Pressure 0.05 hPa
Flash Point 91 °C
Refractive Index 1.4393 (28 °C, 589 nm)
Density 1.072 g/cm3 (20 °C)
Synonyms Cyanoacetic acid allyl ester
Hill Formula C6H7NO2
Chemical Formula NCCH2COOCH2CH=CH2

 

ORDER

SPECIFICATIONS

RELATED INFORMATION

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your basket, enter a quantity and click Add to Basket.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty