Keep my session open?
Ending In 
The session is expired
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Update to Avantor’s response to the coronavirus (COVID-19) pandemic

Butyl phosphate (mixture of mono and diesters) for synthesis, Sigma-Aldrich®

Supplier: EMD MILLIPORE - AA – CORP
Danger

M-Clarity™ quality level = MQ200

8.20251.0100 8.20251.1000
EM8.20251.0100CS 710 USD InStock
EM8.20251.0100 EM8.20251.1000
Butyl phosphate (mixture of mono and diesters) for synthesis, Sigma-Aldrich®
Butyl phosphate (mixture of mono and diesters)


CAS Number: 12788-93-1
UN: 1718
ADR: 8,III
Order Now
Specification Test Results
Solubility (20 °C) partly soluble
Boiling Point >145 °C (1013 hPa)
Flash Point >108.5 °C
Density 1.13 g/cm3 (20 °C)
Synonyms Dibutyl phosphate, Phosphoric acid butyl ester

 

ORDER

SPECIFICATIONS

RELATED INFORMATION

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your basket, enter a quantity and click Add to Basket.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty