Keep my session open?
Ending In 
The session is expired
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Update to Avantor’s response to the coronavirus (COVID-19) pandemic

Guanidinium carbonate for synthesis, Sigma-Aldrich®

Supplier: EMD MILLIPORE - AA – CORP
Danger

M-Clarity™ quality level = MQ200

8.20611.0100 8.20611.1000
EM8.20611.0100CS 8442 USD InStock
EM8.20611.0100 EM8.20611.1000
Guanidinium carbonate for synthesis, Sigma-Aldrich®
Guanidinium carbonate


Formula: C₃H₁₂N₆O₃
MW: 180.17 g/mol
Melting Pt: 197
MDL Number: MFCD00013029
CAS Number: 593-85-1
Order Now
Specification Test Results
Solubility 500 g/l (30 °C)
Melting Point 197 °C
Molar Mass 180.17 g/mol
Bulk Density 500 - 700 kg/m3
Density 1.25 g/cm3 (20 °C)
pH 11.5 (110 g/l, H2O, 20 °C)
Ignition Point 530 °C
Hill Formula C3H12N6O3
Chemical Formula (CH5N3)2 * H2CO3

 

ORDER

SPECIFICATIONS

RELATED INFORMATION

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your basket, enter a quantity and click Add to Basket.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty