Keep my session open?
Ending In 
The session is expired
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Update to Avantor’s response to the coronavirus (COVID-19) pandemic

2-Ethyl-9,10-anthraquinone for synthesis, Sigma-Aldrich®

Supplier: EMD MILLIPORE - AA – CORP
Warning

M-Clarity™ quality level = MQ200

8.21934.0100 8.21934.0250
EM8.21934.0100CS 863 USD InStock
EM8.21934.0100 EM8.21934.0250
2-Ethyl-9,10-anthraquinone for synthesis, Sigma-Aldrich®
2-Ethyl-9,10-anthraquinone


Formula: C₁₆H₁₂O₂
MDL Number: MFCD00001237
CAS Number: 84-51-5
UN: 3077
ADR: 9,III
Order Now
Specification Test Results
Solubility 0.0007 g/l (20 °C)
Melting Point 107 - 110 °C
Molar Mass 236.26 g/mol
Bulk Density 650 kg/m3
Boiling Point 180 - 190 °C
Vapor Pressure 0.00056 hPa (70 °C)
Flash Point >210 °C
Ignition Point 485 °C
Hill Formula C16H12O2

 

ORDER

SPECIFICATIONS

RELATED INFORMATION

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your basket, enter a quantity and click Add to Basket.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty