Keep my session open?
Ending In 
The session is expired
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Update to Avantor’s response to the coronavirus (COVID-19) pandemic

Dimethyl disulphide for synthesis, Sigma-Aldrich®

Supplier: EMD MILLIPORE - AA – CORP
Danger

M-Clarity™ quality level = MQ200

8.20791.0005 8.20791.0250 8.20791.1000 8.20791.9027
EM8.20791.0005CS 2319 USD InStock
EM8.20791.0005 EM8.20791.0250 EM8.20791.1000 EM8.20791.9027
Dimethyl disulphide for synthesis, Sigma-Aldrich®
Dimethyl disulphide


Formula: C₂H₆S₂
MW: 94.2 g/mol
Boiling Pt: 110
Melting Pt: -98
Flash Pt: 16
MDL Number: MFCD00008561
CAS Number: 624-92-0
UN: 2381
ADR: 3 II
Order Now
Specification Test Results
Solubility 2.5 g/l (20 °C)
Melting Point -85 °C
Molar Mass 94.20 g/mol
Boiling Point 110 °C (1013 hPa)
Vapor Pressure 28 hPa (20 °C)
Flash Point 16 °C
Refractive Index 1.525 (20 °C, 589 nm)
Explosion Limit 1.1 - 16.1 %(V)
Density 1.06 g/cm3 (20 °C)
Ignition Point 370 °C
Synonyms Methyl disulfide
Hill Formula C2H6S2
Chemical Formula CH3SSCH3

 

ORDER

SPECIFICATIONS

RELATED INFORMATION

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your basket, enter a quantity and click Add to Basket.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty