Keep my session open?
Ending In 
The session is expired
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Update to Avantor’s response to the coronavirus (COVID-19) pandemic

Dodecylbenzene for synthesis, Sigma-Aldrich®

Supplier: EMD MILLIPORE - AA – CORP

M-Clarity™ quality level = MQ200

8.20990.0025 8.20990.0100
EM8.20990.0025CS 11221 USD InStock
EM8.20990.0025 EM8.20990.0100
Dodecylbenzene for synthesis, Sigma-Aldrich®
Dodecylbenzene


Formula: C₁₈H₃₀
MW: 246.44 g/mol
Boiling Pt: 275
Melting Pt: -4
MDL Number: MFCD00008974
CAS Number: 123-01-3
Order Now
Specification Test Results
Solubility (20 °C) insoluble
Melting Point 3 °C
Molar Mass 246.44 g/mol
Boiling Point 275 - 300 °C (1013 hPa)
Vapor Pressure 0.1 hPa (50 °C)
Flash Point 124 °C
Refractive Index 1.482 (20 °C, 589 nm)
Density 0.85 g/cm3 (20 °C)
Synonyms Dodecyl benzene
Hill Formula C18H30
Chemical Formula C6H5(CH2)11CH3

 

ORDER

SPECIFICATIONS

RELATED INFORMATION

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your basket, enter a quantity and click Add to Basket.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty