Keep my session open?
Ending In 
The session is expired
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Update to Avantor’s response to the coronavirus (COVID-19) pandemic

Propionamide for synthesis, Sigma-Aldrich®

Supplier: EMD MILLIPORE - AA – CORP
Warning

M-Clarity™ quality level = MQ200

8.21046.0050 8.21046.0250
EM8.21046.0050CS 13623 USD InStock
EM8.21046.0050 EM8.21046.0250
Propionamide for synthesis, Sigma-Aldrich®
Propionamide


Formula: C₃H₇NO
MW: 73.09 g/mol
Boiling Pt: 213
Melting Pt: 79
MDL Number: MFCD00008039
CAS Number: 79-05-0
Merck Index: 12,08009
Order Now
Specification Test Results
Solubility (20 °C) soluble
Melting Point 75 - 78 °C
Molar Mass 73.1 g/mol
Boiling Point 213 °C
Hill Formula C3H7NO
Chemical Formula CH3CH2CONH2

 

ORDER

SPECIFICATIONS

RELATED INFORMATION

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your basket, enter a quantity and click Add to Basket.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty