Keep my session open?
Ending In 
The session is expired
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Update to Avantor’s response to the coronavirus (COVID-19) pandemic

1,6-Diisocyanatohexane for synthesis, Sigma-Aldrich®

Supplier: EMD MILLIPORE - AA – CORP
Danger

M-Clarity™ quality level = MQ200

8.22066.0250 8.22066.1000
EM8.22066.0250CS 1404 USD InStock
EM8.22066.0250 EM8.22066.1000
1,6-Diisocyanatohexane for synthesis, Sigma-Aldrich®
1,6-Diisocyanatohexane


Formula: C₈H₁₂N₂O₂
MDL Number: MFCD00002047
CAS Number: 822-06-0
UN: 2281
ADR: 6.1,II
Order Now
Specification Test Results
Solubility (20 °C) (decomposition)
Melting Point -67 °C
Molar Mass 168.2 g/mol
Boiling Point 255 °C (1013 hPa)
Vapor Pressure 0.007 hPa (20 °C)
Flash Point 130 °C
Refractive Index 1.4525 (20 °C, 589 nm)
Explosion Limit 0.9 - 9.5 %(V)
Density 1.05 g/cm3 (25 °C)
Ignition Point 454 °C
Synonyms 1,6-Diisocyanatohexane
Hill Formula C8H12N2O2
Chemical Formula OCN(CH2)6NCO

 

ORDER

SPECIFICATIONS

RELATED INFORMATION

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your basket, enter a quantity and click Add to Basket.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty