Keep my session open?
Ending In 
The session is expired
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Update to Avantor’s response to the coronavirus (COVID-19) pandemic

Benzyl propionate for synthesis, Sigma-Aldrich®

Supplier: EMD MILLIPORE - AA – CORP

M-Clarity™ quality level = MQ200

8.43784.0100 8.43784.0500
EM8.43784.0100CS 742 USD InStock
EM8.43784.0100 EM8.43784.0500
Benzyl propionate for synthesis, Sigma-Aldrich®
Benzyl propionate


Formula: C₁₀H₁₂O₂
MW: 164.2 g/mol
Boiling Pt: 222
MDL Number: MFCD00027003
CAS Number: 122-63-4
Order Now
Specification Test Results
Solubility (20 °C) practically insoluble
Molar Mass 164.21 g/mol
Boiling Point 222 °C (1013 hPa)
Flash Point 96 °C
Density 1.029 - 1.033 g/cm3 (20 °C)
Hill Formula C10H12O2

 

ORDER

SPECIFICATIONS

RELATED INFORMATION

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your basket, enter a quantity and click Add to Basket.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty