Keep my session open?
Ending In 
The session is expired
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Update to Avantor’s response to the coronavirus (COVID-19) pandemic

D-(+)-Cellohexaose ≥95%

Supplier: Origen Biomedical, Inc

J67610-8PL J67610-LB0 J67610-MCR
AAJ67610-8PLEA 210 USD InStock
AAJ67610-8PL AAJ67610-LB0 AAJ67610-MCR
D-(+)-Cellohexaose ≥95%
D-(+)-Cellohexaose
D-(+)-Cellohexaose, 95%

CAS No.: 2478-35-5 Molecular Formula: C36H62O31 Formula Weight: 990.86 Storage Temperature: -30°C to -10°C Physical Form: Powder Sensitivity: Hygroscopic Appearance: White to off-white MDL No.: MFCD00466913


Formula: C₃₆H₆₂O₃₁
Storage Temperature: Freezer
MDL Number: MFCD00466913
CAS Number: 2478-35-5
Order Now
Specification Test Results
Assay Minimum 95

 

ORDER

SPECIFICATIONS

RELATED INFORMATION

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your basket, enter a quantity and click Add to Basket.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty