Keep my session open?
Ending In 
The session is expired
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Update to Avantor’s response to the coronavirus (COVID-19) pandemic

Neodymium triiodide, ultra dry ≥99.9%, powder

Supplier: Origen Biomedical, Inc
Warning

Ampuled under argon

047157-03 047157-06
AA47157-03EA 117 USD InStock
AA47157-03 AA47157-06
Neodymium triiodide, ultra dry ≥99.9%, powder
Neodymium triiodide
Neodymium(III) iodide, ultra dry, 99.9% (metals basis). Powder, Ampouled under argon.

Purity: 99.9% CAS No.: 13813-24-6 Molecular Formula: NdI3 Formula Weight: 524.95 Physical Form: Powder, Ampouled under argon Sensitivity: Moisture and Light Sensitive Appearance: Black Odor: Odorless MDL No.: MFCD00016248


Formula: I₃Nd
MW: 524.95
Melting Pt: 784
Density: 5.85
Storage Temperature: Ambient
MDL Number: MFCD00016248
CAS Number: 13813-24-6
EINECS: 237-467-7
Order Now
Specification Test Results
Assay Minimum 99.9

 

ORDER

SPECIFICATIONS

RELATED INFORMATION

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your basket, enter a quantity and click Add to Basket.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty