Keep my session open?
Ending In 
The session is expired
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Update to Avantor’s response to the coronavirus (COVID-19) pandemic

H-D-Lys(Boc)-OH 98%

Supplier: Origen Biomedical, Inc

H62736-03 H62736-MD
AAH62736-03EA 131 USD InStock
AAH62736-03 AAH62736-MD
H-D-Lys(Boc)-OH 98%
H-D-Lys(Boc)-OH
N^e-Boc-D-lysine, 98%

Purity: 98% CAS No.: 31202-69-4 Molecular Formula: C11H22N2O4 Formula Weight: 246.30 Storage Temperature: -30°C to -10°C Physical Form: Powder Appearance: White MDL No.: MFCD00076958


Formula: C₁₁H₂₂N₂O₄
MW: 246.307 g/mol
Storage Temperature: Refrigerator
MDL Number: MFCD00076958
CAS Number: 31202-69-4
Order Now
Specification Test Results
Assay Minimum 98

 

ORDER

SPECIFICATIONS

RELATED INFORMATION

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your basket, enter a quantity and click Add to Basket.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty