Keep my session open?
Ending In 
The session is expired
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Update to Avantor’s response to the coronavirus (COVID-19) pandemic

5-(Trifluoromethyl)picolinonitrile 98%

Supplier: Origen Biomedical, Inc
Warning

H64486-03 H64486-06
AAH64486-03EA 37.5 USD InStock
AAH64486-03 AAH64486-06 AAH64486-MD
5-(Trifluoromethyl)picolinonitrile 98%
5-(Trifluoromethyl)picolinonitrile
2-Cyano-5-(trifluoromethyl)pyridine, 98%

Purity: 98% CAS No.: 95727-86-9 Molecular Formula: C7H3F3N2 Formula Weight: 172.11 Physical Form: Solid Beilstein Reference: 13339826 UN No.: 3439 MDL No.: MFCD01001119


Formula: C₇H₃F₃N₂
MW: 172.11 g/mol
Storage Temperature: Ambient
MDL Number: MFCD01001119
CAS Number: 95727-86-9
UN: 3439
ADR: 6.1 III
Order Now
Specification Test Results
Assay Minimum 98

 

ORDER

SPECIFICATIONS

RELATED INFORMATION

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your basket, enter a quantity and click Add to Basket.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty