H65365-06
H65365-14
AAH65365-06EA
56.5
USD
InStock
AAH65365-06
AAH65365-14
AAH65365-22
β-Alaninamide hydrochloride 96%
beta-Alaninamide hydrochloride
^b-Alaninamide hydrochloride, 96%
Purity: 96%
CAS No.: 64017-81-8
Molecular Formula: C3H8N2O.HCl
Formula Weight: 124.57
Storage Temperature: 2°C to 8°C
Physical Form: Solid
Sensitivity: Air Sensitive
Melting Point: 149-154°
Beilstein Reference: 3909917
MDL No.: MFCD00060747