Keep my session open?
Ending In 
The session is expired
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Update to Avantor’s response to the coronavirus (COVID-19) pandemic

(S)-tert-Butyl 5-oxopyrrolidine-2-carboxylate 95%

Supplier: Origen Biomedical, Inc
Warning

H65385-06 H65385-14
AAH65385-06EA 62.7 USD InStock
AAH65385-06 AAH65385-14
(S)-tert-Butyl 5-oxopyrrolidine-2-carboxylate 95%
(S)-tert-Butyl-5-oxopyrrolidine-2-carboxylate
tert-Butyl (S)-5-oxopyrrolidine-2-carboxylate, 95%

Purity: 95% CAS No.: 35418-16-7 Molecular Formula: C9H15NO3 Formula Weight: 185.22 Physical Form: Powder Appearance: White Beilstein Reference: 3590226 MDL No.: MFCD06659481


Formula: C9H15NO3
MW: 185.22 g/mol
Storage Temperature: Ambient
MDL Number: MFCD06659481
CAS Number: 35418-16-7
EINECS: 252-555-5
Order Now
Specification Test Results
Assay Minimum 95

 

ORDER

SPECIFICATIONS

RELATED INFORMATION

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your basket, enter a quantity and click Add to Basket.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty