Keep my session open?
Ending In 
The session is expired
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Update to Avantor’s response to the coronavirus (COVID-19) pandemic

beta-Alanyl-L-Histidine

Supplier: Origen Biomedical, Inc

H65668-06 H65668-14
AAH65668-06EA 58.4 USD InStock
AAH65668-06 AAH65668-14
beta-Alanyl-L-Histidine
Proteins and Peptides
^b-Alanyl-L-histidine, 95%

  • EINECS: 206-169-9
  • Melting point: 253 °C

Purity: 95% CAS No.: 305-84-0 Molecular Formula: C9H14N4O3 Formula Weight: 226.24 Storage Temperature: -30°C to -10°C Physical Form: Crystalline Appearance: White Beilstein Reference: 87671 MDL No.: MFCD00005207


Order Now

 

ORDER

SPECIFICATIONS

RELATED INFORMATION

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your basket, enter a quantity and click Add to Basket.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty