H65792-03
H65792-06
H65792-MD
AAH65792-03EA
89.4
USD
InStock
AAH65792-03
AAH65792-06
AAH65792-MD
H-Asn-OtBu·HCl 95%
H-Asn-OtBu·HCl
L-Asparagine tert-butyl ester hydrochloride, 95%
Purity: 95%
CAS No.: 63094-81-5
Molecular Formula: C8H16N2O3.HCl
Formula Weight: 224.69
Physical Form: Powder
Appearance: White
Beilstein Reference: 6585123
Optical Rotation: +11°(c=1 in methanol)
MDL No.: MFCD00039079