H66090-03
H66090-06
AAH66090-03EA
44.6
USD
InStock
AAH66090-03
AAH66090-06
AAH66090-MD
6-Nitroanthranilic acid 97%
6-Nitroanthranilic acid
2-Amino-6-nitrobenzoic acid, 97%
Purity: 97%
CAS No.: 50573-74-5
Molecular Formula: C7H6N2O4
Formula Weight: 182.13
Storage Temperature: -30°C to -10°C
Physical Form: Solid
Beilstein Reference: 2807130
MDL No.: MFCD00034785