H66189-03
H66189-06
AAH66189-03EA
68
USD
InStock
AAH66189-03
AAH66189-06
AAH66189-MD
Fmoc-L-homocitrulline 95%
Fmoc-L-homocitrulline
N^a-Fmoc-N^e-carbamoyl-L-lysine, 95%
Purity: 95%
CAS No.: 201485-17-8
Molecular Formula: C22H25N3O5
Formula Weight: 411.46
Storage Temperature: -30°C to -10°C
Physical Form: Solid
MDL No.: MFCD00237387