Keep my session open?
Ending In 
The session is expired
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Update to Avantor’s response to the coronavirus (COVID-19) pandemic

Fmoc-Asp(Ompe)-OH 98% (by HPLC)

Supplier: Origen Biomedical, Inc

H66281-03 H66281-06
AAH66281-03EA 76.99 USD InStock
AAH66281-03 AAH66281-06 AAH66281-MD
Fmoc-Asp(Ompe)-OH 98% (by HPLC)
Fmoc-Asp(Ompe)-OH
N-Fmoc-L-aspartic acid 4-methylpentyl ester, 98%

Purity: 98% CAS No.: 180675-08-5 Molecular Formula: C25H29NO6 Formula Weight: 439.51 Storage Temperature: -30°C to -10°C Physical Form: Solid MDL No.: MFCD10001333


Formula: C₂₅H₂₉NO₆
MW: 439.508 g/mol
Storage Temperature: Refrigerator
MDL Number: MFCD10001333
CAS Number: 180675-08-5
Order Now
Specification Test Results
Assay (HPLC) >97.5%

 

ORDER

SPECIFICATIONS

RELATED INFORMATION

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your basket, enter a quantity and click Add to Basket.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty