H66390-06
H66390-14
AAH66390-06EA
68
USD
InStock
AAH66390-06
AAH66390-14
4-[(tert-Butoxycarbonylamino)methyl]benzoic acid 97%
4-[(tert-Butoxycarbonylamino)methyl]benzoic acid
4-(Boc-aminomethyl)benzoic acid, 98%
Purity: 98%
CAS No.: 33233-67-9
Molecular Formula: C13H17NO4
Formula Weight: 251.28
Physical Form: Crystalline
Appearance: Off-white
Melting Point: 164-168°
Beilstein Reference: 2854286
MDL No.: MFCD00228182