Keep my session open?
Ending In 
The session is expired
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Update to Avantor’s response to the coronavirus (COVID-19) pandemic

Fmoc-N-Me-Phe-OH 95%

Supplier: Origen Biomedical, Inc

H66565-03 H66565-06
AAH66565-03EA 59.5 USD InStock
AAH66565-03 AAH66565-06
Fmoc-N-Me-Phe-OH 95%
Fmoc-N-Me-Phe-OH
N-Fmoc-N-methyl-L-phenylalanine, 95%

Purity: 95% CAS No.: 77128-73-5 Molecular Formula: C25H23NO4 Formula Weight: 401.46 Storage Temperature: 2°C to 8°C Physical Form: Powder Appearance: Off-white Beilstein Reference: 4768138 MDL No.: MFCD00151938


Formula: C₂₅H₂₃NO₄
MW: 401.462 g/mol
Storage Temperature: Ambient
MDL Number: MFCD00151938
CAS Number: 77128-73-5
Order Now
Specification Test Results
Assay Minimum 95

 

ORDER

SPECIFICATIONS

RELATED INFORMATION

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your basket, enter a quantity and click Add to Basket.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty