Vacuum Membrane Filter Units, Advantec MFS
Supplier: Advantec MFS
311760 311700 311850 311860 311750 311800
101105-124PK 1882.71 USD InStock
101105-124 101105-162 101105-056 101104-886 101105-012 101105-476
Vacuum Membrane Filter Units, Advantec MFS
Funnels Membrane Filter Funnels
Vaccum membrane filtration unit collect aliquots of filtrate during the filtration process using the 2-way Y bore PTFE valve.
It prevents venting of hazardous sample or solvents to atmosphere, and collects multiple samples with additional receiver sample tubes. Features two mounting options, such as P/FC 1 mounts on standard manifold or 1 L flask using a #8 stopper and P/FC 2 has female 40/35 joint to mount on an extraction manifold or 1 L flask.
Información para pedidos: Available with sintered glass, stainless steel, or PTFE support.
Order Now
ORDER
SPECIFICATIONS
ACCESSORIES
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your basket, enter a quantity and click Add to Basket.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty