Human Recombinant Follistatin (from E. coli)
Supplier: FUJIFILM IRVINE SCIENTIFIC, INC.
100-41-100UG 100-41-1MG 100-41-20UG 100-41-5UG 100-41-500UG
10787-578EA 866.35 USD InStock
10787-578 10789-032 10789-034 10789-036 10821-982
Human Recombinant Follistatin (from E. coli)
Proteins and Peptides
Follistatin is an autocrine, activin-binding protein that is ubiquitously expressed with highest expression levels being in the ovary and skin. Follistatin negatively regulates the signaling of transforming growth factor beta (TGF-β) family members, such as activin, bone morphogenetic proteins (BMPs), myostatin, growth differentiation factor 11 (GDF-11), and TGF-β1. Follistatin functions as an antagonist by binding TGF-β family members to block interaction with their signaling receptors. Follistatin also inhibits the secretion of follicle-stimulating hormone (FSH) from the anterior pituitary.
  • High quality
  • Low endotoxin
  • Affordable
Precaución: For research use only.
Order Now
ORDER
SPECIFICATIONS
RELATED INFORMATION
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your basket, enter a quantity and click Add to Basket.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty