Human Recombinant IL-35 (from HEK293 cells)
Supplier: FUJIFILM IRVINE SCIENTIFIC, INC.
100-49-100UG 100-49-10UG
10787-646EA 2618.82 USD InStock
10787-646 10787-648 10821-838
Human Recombinant IL-35 (from HEK293 cells)
Proteins and Peptides
Interleukin 35 (IL-35) is a member of the IL-12 cytokine family and is produced by regulatory T cells (Tregs). IL-35 is a heterodimeric cytokine that is comprised of the p35 subunit (IL-12A) and the Epstein-Barr virus induced gene 3 subunit (EBI3/IL-27B). IL-35 binds the IL-12Rbeta2/gp130 hetero- and homodimers to activate STAT1 and STAT4 signaling.IL-35 functions as a suppressor of immune cell inflammatory responses.
  • High quality
  • Low endotoxin
  • Affordable
Precaución: For research use only.
Order Now
ORDER
SPECIFICATIONS
RELATED INFORMATION
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your basket, enter a quantity and click Add to Basket.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty