Keep my session open?
Ending In 
The session is expired
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Update to Avantor’s response to the coronavirus (COVID-19) pandemic

Ethyl-2-chloro-3-pyridylglyoxylate ≥97%

Supplier: BEANTOWN CHEMICAL - BTC

121020-1G 121020-5G
BT121020-1GEA 205 USD InStock
BT121020-1G BT121020-5G
Ethyl-2-chloro-3-pyridylglyoxylate ≥97%
Ethyl-2-chloro-3-pyridylglyoxylate
CAS: 902837-56-3; MDL No: MFCD07781194 Liquid; Molecular Formula: C9H8ClNO3; MW: 213.62

  • Liquid


Formula: C₉H₈ClNO₃
MDL Number: MFCD07781194
CAS Number: 902837-56-3
Order Now
Specification Test Results
Assay 97%

 

ORDER

SPECIFICATIONS

RELATED INFORMATION

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your basket, enter a quantity and click Add to Basket.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty